Skip to Content
Merck
All Photos(1)

Key Documents

N12249

Isofenphos

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24NO4PS
CAS Number:
Molecular Weight:
345.39
Beilstein:
2949615
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

form

liquid

packaging

ampule of 100 mg

manufacturer/tradename

Chem Service, Inc. PS-1003

SMILES string

CCOP(=S)(NC(C)C)Oc1ccccc1C(=O)OC(C)C

InChI

1S/C15H24NO4PS/c1-6-18-21(22,16-11(2)3)20-14-10-8-7-9-13(14)15(17)19-12(4)5/h7-12H,6H2,1-5H3,(H,16,22)

InChI key

HOQADATXFBOEGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isofenphos is an organophosphorus pesticide, used for the control of larvae of Popillia japonica Newman, soil-dwelling pests of corn (Amaze), Delia antiqua (Onion flies), Chamaepsila rosae (carrot flies), wireworms, etc. Toxicologically, the pesticide acts by the inhibition of acetylcholinesterase activity.[1][2][3]

Application

Isofenphos may be used as an analytical reference standard for the determination of the pesticide residue in meat samples,[4] leeks,[5] rice[6] and human tissues[7] by various chromatography techniques.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monoclonal antibody-based enzyme-linked immunosorbent assays for the detection of the organophosphorus insecticide isofenphos
Lee WY, et al.
Analytica Chimica Acta, 557(1-2), 169-178 (2006)
Dong Woon Lee et al.
Journal of economic entomology, 95(5), 918-926 (2002-10-31)
The oriental beetle Exomala (Anomala) orientalis (Waterhouse) is an important pest of turfgrass in Korean golf courses, and although a few chemical insecticides are registered for insect pest control, they are not very effective against scarab larvae. There is also
Rapid determination of organophosphorous pesticides in leeks by gas chromatography-triple quadrupole mass spectrometry
Qu LJ, et al.
Food Chemistry, 122(1), 327-332 (2010)
A Garrido-Frenich et al.
Journal of chromatography. A, 1133(1-2), 315-321 (2006-09-05)
The assessment of the recovery factor with the analyte concentration in meat samples has been studied for the determination of organochlorine and organophosphorus pesticides in meat by gas chromatography-triple quadrupole mass spectrometry (GC-MS/MS). For that purpose, recent IUPAC recommendations, which
E E Calore et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 51(1), 27-33 (1999-02-27)
The expression of sarcoplasmic esterases, lipases as well as the lipid content in the myofibers of the diaphragm of rats intoxicated with the organophosphate isofenphos was studied. Lipid accumulation was documented at light, electron microsopic and by morphometric studies. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service