G1007511
UHPLC Analysis of Orthoclone OKT3 IgG2 Monoclonal Antibody on BIOshell A400 Protein C4
application for UHPLC
About This Item
technique(s)
UHPLC: suitable
test parameters
column: BIOshell A400 Protein C4; 10 cm x 2.1 mm I.D., 3.4 μm particles (66825-U)
mobile phase: [A] 90:10 (0.1% TFA in water): (0.1% TFA in acetonitrile); [B] 10:90 (0.1% TFA in water): (0.1% TFA in acetonitrile)
gradient: 0% B to 50%B in 50 min; 50%B to 100% B in 10 min
flow rate: 0.2 mL/min
pressure: 1650 psi (114 bar)
column temp.: 75 °C
detector: UV, 215 nm
injection: 1.0 μL
sample: Orthoclone OKT3, 1 mg/mL, 0.05% TFA in water
suitability
application for UHPLC
application(s)
life science and biopharma
Analysis Note
Legal Information
mobile phase component
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service