Skip to Content
Merck
All Photos(1)

Documents

G006125

HPLC Analysis of Proteins by Size Exclusion on Zenix® Columns, Effect of Pore Size

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Zenix SEC-150, 30 cm x 7.8 mm I.D., 3 μm, 150 Å particles (Z777018)
column: Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 μm, 300 Å particles (Z777033)
mobile phase: 150 mM sodium phosphate, pH 7
flow rate: 1 mL/min
column temp.: ambient
detector: UV, 214 nm
injection: 10 μL
sample: each 1 mg/mL except uracil, 2.5 mg/mL

suitability

application for HPLC

application(s)

life science and biopharma
pharmaceutical (small molecule)

Legal Information

Zenix is a registered trademark of Sepax Technologies

Analyte

Description

    Thyroglobulin from bovine thyroid, powder, ≥90% (agarose gel electrophoresis)
    Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)
    Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
    Uracil, ≥99.0% (T)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service