Skip to Content
Merck
All Photos(1)

Key Documents

G005946

HPLC Analysis of Proteins on Antibodix® WCX-NP10, Comparison of WCX Columns

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Antibodix WCX-NP10, 25 cm x 4.6 mm I.D., 10 μm particles (Z777272)
mobile phase: (A) 10 mM sodium phosphate buffer, pH 6.0; (B) A + 1.0 M NaCl
gradient: 0 to 100% B in 42 min
flow rate: 1.0 mL/min
column temp.: ambient
detector: UV, 214 nm
injection: 5 μL
sample: 1 mg/mL each

suitability

application for HPLC

application(s)

life science and biopharma
pharmaceutical (small molecule)

Legal Information

Antibodix is a registered trademark of Sepax Technologies

Analyte

Description

    Worthington Accessories, cap for 4 L Dewar
    Lysozyme from chicken egg white, powder or granules, ≥39,000 units/mg protein
    Ribonuclease A from bovine pancreas, ≥70 Kunitz units/mg protein, lyophilized

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service