Skip to Content
Merck
All Photos(1)

Documents

G005817

LC/MS (TOF) Analysis of Antiarrhythmic Drugs and Metabolites on Ascentis® Express HILIC

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express HILIC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase: [A] 5 mM ammonium formate; [B] 5 mM ammonium formate in acetonitrile; (5:95, A:B, pH 7.0 with formic acid)
flow rate: 0.4 mL/min
column temp.: 35 °C
detector: ESI(+), full scan, m/z 200-800
injection: 0.5 μL
sample: each compound, 300 ng/mL in 1% formic acid acetonitrile:water, 75:25

suitability

application for HPLC

application(s)

clinical

Analysis Note

The basic nature of these compounds makes them targets for HILIC chromatographic separation. HILIC mobile phases consist of a high composition of acetonitrile, which facilitates the direct analysis of precipitated plasma samples without the need for additional sample solvent exchange. In most cases, the high organic mobile phase also facilitates increased analyte response with ESI(+) MS detection. Separation was performed on an Ascentis Express HILIC Fused-Core HPLC column. High purity solvents provided clean, robust operation. Cerilliant CRMs provided reliable quantification.

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Amiodarone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    N-Desethylamiodarone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
    Lidocaine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
    (±)-Flecainide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service