Skip to Content
Merck
All Photos(1)

Documents

G005801

GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on SUPELCOWAX 10 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis

suitable for GC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable

test parameters

column: SUPELCOWAX 10, 15 m x 0.10 mm I.D., 0.10 μm (24343)
oven: 80°C (0.3 min), 30 °C/min to 180 °C (0.5 min), 10 °C/min to 200 °C (1 min)
inj. temp.: 250 °C
detector: FID, 250 °C
carrier gas: hydrogen, 45 cm/sec, constant
injection: 0.1 μL 200:1 split
liner: 2 mm I.D., straight
sample: bacterial methyl ester standards in methyl caproate, 10 mg/mL (47080-U)

application(s)

clinical

Analysis Note

Food-borne microbial pathogens can cause illness in humans. Because each bacterium has a unique cellular fatty acid profile, GC analysis can be used to help with identification. Analysis is performed following derivatization of cellular fatty acids to methyl esters. These are called bacterial acid methyl esters (BAMEs) instead of fatty acid methyl esters (FAMEs) to signify the bacterial source. The chromatogram shown is the analysis of a standard mix containing 26 BAME compounds. The polar nature of the column, along with the use of Fast GC column dimensions and instrument conditions, allows good peak shape and resolution in just 7 minutes.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service