Skip to Content
Merck
All Photos(1)

Key Documents

ES5578

ChromegaChiral Chiral CCO F4, 3 Micron HPLC Column

3 μm particle size, L × I.D. 10 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$4,440.00

$4,440.00


Estimated to ship on14 April 2025



Select a Size

Change View
1 EA
$4,440.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

$4,440.00


Estimated to ship on14 April 2025


material

stainless steel column

Quality Level

product line

ChromegaChiral

manufacturer/tradename

ES Industries 125151-CCO-F4

packaging

1 ea of

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

10 cm × 4.6 mm

matrix

fully porous particle

particle size

3 μm

pore size

1000 Å pore size

operating pH

2-8

separation technique

chiral
supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

ChromegaChiral CCO F4 is a 4-Fluoro 3-methylphenyl cellulose phase which can be used in SFC or HPLC. The addition of a fluorine atom into a phenyl carbamate cellulose structure is useful in promoting a fluorophilic retention mechanism which provides improved retention for fluorinated compounds. A fluorophilic retention mechanism is particularly useful in medicinal chemistry and drug discovery, where more than a third of newly approved small molecule drugs contain fluorine.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChromegaChiral is a trademark of ES Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service