Skip to Content
Merck
All Photos(1)

Key Documents

ES5554

ChromegaChiral Chiral CCO F2, 5 Micron HPLC Column

5 μm particle size, L × I.D. 15 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$3,840.00

$3,840.00


Check Cart for Availability


Select a Size

Change View
1 EA
$3,840.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

$3,840.00


Check Cart for Availability

material

stainless steel column

Quality Level

product line

ChromegaChiral

manufacturer/tradename

ES Industries 135251-CCO-F2

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

fully porous particle

matrix active group

2-fluoro 5-methylphenyl cellulose phase

particle size

5 μm

pore size

1000 Å pore size

operating pH

2-8

separation technique

chiral
supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

ChromegaChiral CCO F2 is a 2-Fluoro 5-methylphenyl cellulose phase which can be used in SFC or HPLC. The addition of a fluorine atom into a phenyl carbamate cellulose structure can be useful in promoting a fluorophilic retention mechanism which can provide improved retention for fluorinated compounds. A fluorophilic retention mechanism can be particularly useful in medicinal chemistry and drug discovery, where more than a third of newly approved small molecule drugs contain fluorine.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChromegaChiral is a trademark of ES Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service