Skip to Content
Merck
All Photos(1)

Key Documents

ES5074

GreenSep 4-Ethyl Pyridine SFC 3 Micron HPLC Column

3 μm particle size, L × I.D. 5 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.52
L × I.D.:
5 cm × 4.6 mm
particle size:
3 μm
matrix active group:
4-ethyl pyridine phase
pore size:
120 Å pore size
matrix:
fully porous particle

material

stainless steel column

product line

GreenSep

manufacturer/tradename

ES Industries 115191-GS-PYE4

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

5 cm × 4.6 mm

matrix

fully porous particle

matrix active group

4-ethyl pyridine phase

particle size

3 μm

pore size

120 Å pore size

operating pH

2-8

separation technique

supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

GreenSep 4-Ethyl Pyridine is an alternative to and provides different selectivity to GreenSep Ethyl Pyridine (2- ethyl pyridine). The chromatogram below shows the separation of chemicals such as ibuprofen, ketoprofen and niflumic acid, without peak shape modifiers, and is a prime example of the superior peak shape performance obtainable with the GreenSep 4-Ethyl Pyridine column.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service