Skip to Content
Merck
All Photos(1)

Key Documents

ERMBF443D

Soya bean GMB151

nominal 1% GMO, ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$222.00

$222.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$222.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$222.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

Agency

ERM®

quality

nominal 1% GMO

manufacturer/tradename

JRC

application(s)

food and beverages

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please click on the below product number:
ERMBF443D

Legal Information

ERM is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service