ERMBD286
Paprika Powder (aflatoxin B1, G1)
ERM®, certified reference material
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
85151701
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
Agency
ERM®
manufacturer/tradename
JRC
application(s)
general analytical
storage temp.
−20°C
Related Categories
Analysis Note
For more information please see:
ERMBD286
ERMBD286
Other Notes
Certified for the analytes listed below. See certificate for values and more details.
Biotoxins / Mycotoxins
Aflatoxin B1, Aflatoxin G1
Matrix Group: Spices
Biotoxins / Mycotoxins
Aflatoxin B1, Aflatoxin G1
Matrix Group: Spices
Legal Information
ERM is a registered trademark of European Commission
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service