Skip to Content
Merck
All Photos(1)

Documents

D4294GOSUC

Sulfur in synthetic diesel, ASTM D4294

Calibration Set C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

petroleum

format

matrix material

Looking for similar products? Visit Product Comparison Guide

Components

Set C
1.0000 % (10000ppm), 2.0000 % (20000ppm), 3.0000 % (30000ppm), 4.0000 % (40000ppm), 5.0000 % (50000ppm) . 100 mL ea.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

170.6 °F

Flash Point(C)

77 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service