Skip to Content
Merck
All Photos(1)

Key Documents

CRMUCOCLOW

Flash Point Standard

COC ASTM D 92 (85.0 °C)

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 80 ML
$742.00

$742.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
3 X 80 ML
$742.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$742.00


Check Cart for Availability

Request a Bulk Order

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

168.8 °F

Flash Point(C)

76 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service