Skip to Content
Merck
All Photos(1)

Key Documents

CRM67622

Supelco

Tralomethrin

mixture of stereoisomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(S)-α-Cyano-3-phenoxybenzyl (1R,3S)-2,2-dimethyl-3-[(RS)-1,2,2,2-tetrabromoethyl]cyclopropanecarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$163.00

$163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$163.00

About This Item

Empirical Formula (Hill Notation):
C22H19Br4NO3
CAS Number:
Molecular Weight:
665.01
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$163.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

O=C(OC(C#N)C1=CC=CC(OC2=CC=CC=C2)=C1)C3C(C(Br)C(Br)(Br)Br)C3(C)C

InChI

1S/C22H19Br4NO3/c1-21(2)17(19(23)22(24,25)26)18(21)20(28)30-16(12-27)13-7-6-10-15(11-13)29-14-8-4-3-5-9-14/h3-11,16-19H,1-2H3

InChI key

YWSCPYYRJXKUDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Tralomethrin is a synthetic Type II pyrethroid insecticide that contains an a-cyano group and shows more neurotoxicity than Type I insecticides. It acts by modifying the kinetics of the voltage-gated sodium channels present in the axonal membranes of neurons. In addition, the voltage-gated calcium and chloride channels and GABA (gamma amino butyric acid) receptors are the secondary target sites for tralomethrin.
Tralomethrin is not approved for use in the European Union (EU) as per the Commission Regulation (EC) No 2076/2002 of 20th November 2002 as a plant protection agent under Regulation (EC) No. 1107/2009, repealing the directive 91/414/EEC. But a default maximum residue limit (MRL) of 0.01 mg/kg is applicable for its presence in products of plant origin according to Art 18(1)(b) Reg 396 / 2005.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The tralomethrin CRM may also be used as below:

  • Develop and validate a method relying on the use of a vibrational shaker and QuEChERS-based extraction, accompanied by LC-MS/MS detection to detect 22 antiparasitic drug residues in 36 fish tissue samples
  • Analysis of fat obtained from different animals for the presence of 1 pyrethrin and 18 pyrethroid residues by UHPLC-MS/MS
  • Multi-residue analysis of 379 pesticides in human serum by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Quantification of 19 pyrethroid residues in 262 tea and 209 peeled orange samples by liquid chromatography-electrospray ionization-triple quadrupole mass spectrometry (LC-ESI-QqQ-MS/MS) and their detection by liquid chromatography-electrospray ionization-quadrupole-time-of-flight mass spectrometry (LC-ESI-Q-ToF/MS)
  • Simultaneous determination of 12 pyrethroid insecticide residues in water, sediment, and milk samples by gas chromatography coupled with tandem mass spectrometry (GC/MS/MS) in two different ionization modes of electron ionization (EI) and negative chemical ionization (NCI)
  • Multi-residue determination of 17 pesticide residues in brown rice, onion, and spinach samples by supercritical fluid chromatography-tandem mass spectrometry (SFC-MS/MS)

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L Feo et al.
Rapid communications in mass spectrometry : RCM, 25(7), 869-876 (2011-03-19)
The performance of gas chromatography coupled with tandem mass spectrometry (GC/MS/MS) was tested for the simultaneous determination of twelve pyrethroid insecticides. First, a comparison of two different ionization modes, electron ionization (EI) and negative chemical ionization (NCI), was carried out
Lenise Guimarães de Oliveira et al.
Analytical and bioanalytical chemistry, 411(26), 6913-6929 (2019-08-17)
A method was developed for the analysis of 22 antiparasitic residues belonging to the benzoylurea, organophosphate, pyrimidinamine, pyrethrin and pyrethroid classes in salmon by liquid chromatography coupled with tandem mass spectrometry. Samples were extracted with acetonitrile-water as the extraction solvent
Hui Zhan et al.
Environmental research, 182, 109138-109138 (2020-02-20)
The significant applications of pyrethroid insecticides in agro-ecosystem and household environments have raised serious environmental concerns. Environmental bioremediation has emerged as an effective and eco-friendly approach to remove or neutralize hazardous compounds. Bioaugmentation accelerates pyrethroid degradation in liquid cultures and
Megumi Ishibashi et al.
Journal of chromatography. A, 1266, 143-148 (2012-10-30)
Combination techniques such as gas chromatography/mass spectrometry (GC/MS) and liquid chromatography/mass spectrometry (LC/MS) are commonly used for pesticide residue analysis, but there is no reported method for the simultaneous analysis of multiple pesticides in a sample using a single instrument.
Comprehensive target analysis for 19 pyrethroids in tea and orange samples based on LC-ESI-QqQ-MS/MS and LC-ESI-Q-ToF/MS
Wan J, et al.
LWT--Food Science and Technology, 151, 112072-112072 (2021)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service