Skip to Content
Merck
All Photos(1)

Key Documents

CRM4M7341

Supelco

Acetone- 2,4-DNPH solution

certified reference material, 1000 μg/mL in acetonitrile, ampule of 5 × 1 mL

Synonym(s):

Acetone-2,4-dinitrophenylhydrazone solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$421.00

$421.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 PKG
$421.00

About This Item

Empirical Formula (Hill Notation):
C9H10N4O4
CAS Number:
Molecular Weight:
238.20
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24

$421.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 5 × 1 mL

concentration

1000 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C9H10N4O4/c1-6(2)10-11-8-4-3-7(12(14)15)5-9(8)13(16)17/h3-5,11H,1-2H3

InChI key

YGIXYAIGWMAGIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is 1000 ug/mL the concentration of acetone or acetone-DNPH derivative?

    1 answer
    1. The reported concentration of 1000 µg/mL is in reference to the Acetone- 2,4-DNPH.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service