Skip to Content
Merck
All Photos(1)

Key Documents

CRM47904

Supelco

EPA 501/601 Trihalomethanes Calibration Mix

certified reference material, TraceCERT®, 100 μg/mL each component in methanol, ampule of 1 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$59.30

$59.30


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 PKG
$59.30

About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

$59.30


Available to ship on22 April 2025Details


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 601,501.3
suitable for EPA 601

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

100 μg/mL each component in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Bromodichloromethane
    Bromoform
    Chloroform
    Dibromochloromethane

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service