Skip to Content
Merck
All Photos(1)

Key Documents

CRM47177

Supelco

Formaldehyde-DNPH solution

certified reference material, TraceCERT®, 100 μg/mL in acetonitrile (as aldehyde), ampule of 1 mL

Synonym(s):

Formaldehyde-2,4-DNPH Solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$54.40

$54.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 PKG
$54.40

About This Item

Linear Formula:
H2C=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
210.15
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$54.40


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

Agency

CARB

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

100 μg/mL in acetonitrile (as aldehyde)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1c(ccc(c1)[N+](=O)[O-])NN=C

InChI

1S/C7H6N4O4/c1-8-9-6-3-2-5(10(12)13)4-7(6)11(14)15/h2-4,9H,1H2

InChI key

UEQLSLWCHGLSML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ikuo Ueta et al.
Analytical and bioanalytical chemistry, 407(3), 899-905 (2014-09-10)
A novel miniaturized sample extraction capillary was developed to provide a simple and sensitive method for analyzing gaseous formaldehyde (FA) using conventional high-performance liquid chromatography (HPLC). The extraction capillary was prepared by packing silica gel particles in a stainless steel

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. 저장 조건이 argon gas로 되어있는데 어떤 방식으로 보관을 하는걸까요? 일반 room temperature에서 보관하면 문제가 될까요?

    1 answer
    1. This product should be stored between 2-8°C under an inert gas such as argon.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service