Skip to Content
Merck
All Photos(1)

Key Documents

CRM45544

Supelco

Jodfenphos

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$138.00

$138.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$138.00

About This Item

Empirical Formula (Hill Notation):
C8H8Cl2IO3PS
CAS Number:
Molecular Weight:
413.00
Beilstein:
1885795
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$138.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

COP(=S)(OC)Oc1cc(Cl)c(I)cc1Cl

InChI

1S/C8H8Cl2IO3PS/c1-12-15(16,13-2)14-8-4-5(9)7(11)3-6(8)10/h3-4H,1-2H3

InChI key

LFVLUOAHQIVABZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.Download your certificate at: http://www.sigma-aldrich.com

Application

Environmental
Jodfenphos is an obsolete insecticide used for the protection of stored products, livestock pests and in public health applications. As of Reg. (EC) No 1107/2009 jodfenphos is not approved for use as plant protection product in the European Union (EU). A default maximum residue level (MRL) of 0.01 mg/kg has been set for jodfenphos according to Art 18(1)(b) Reg 396/2005. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Aquatic Acute 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service