Skip to Content
Merck
All Photos(1)

Key Documents

CRM44896

Supelco

ASTM® D6584 1,2,4-Butanetriol Solution

certified reference material, 1000 μg/mL in pyridine, ampule of 5 mL

Synonym(s):

Biodiesel impurity, Butanetriol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product CRM44896 is not currently sold in your country. Contact Technical Service

grade

certified reference material
TraceCERT®

Quality Level

Agency

ASTM® D6584

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 5 mL

concentration

1000 μg/mL in pyridine

technique(s)

gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-8°C

SMILES string

O[C@H](CO)CCO

InChI

1S/C4H10O3/c5-2-1-4(7)3-6/h4-7H,1-3H2/t4-/m0/s1

InChI key

ARXKVVRQIIOZGF-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • ASTM D6584 in Biofuel Production: ASTM D6584 1,2,4-Butanetriol Solution is crucial for enhancing biofuel production techniques, particularly in the esterification processes necessary for converting glycerol into value-added biodiesel products. This application emphasizes its importance in turning biowaste into environmentally friendly fuel options (Keogh et al., 2024).

  • Life Cycle Analysis in Waste Management: Utilizing the ASTM D6584 standard, studies such as the environmental impact assessment of glycerol valorization from biodiesel demonstrate the method′s effectiveness in life cycle perspective analyses. This research supports sustainable waste management practices and the optimization of resource recovery (Tomatis et al., 2024).

  • Biotechnological Applications: The 1,2,4-Butanetriol standard is employed in microbial engineering to enhance lipid production in yeast, which is critical for biofuel generation. This showcases the compound′s utility in genetic engineering and metabolic pathway optimization for industrial biotechnology (Jia et al., 2024).

  • Environmental Health Research: In air quality management, ASTM D6584 butanetriol solutions are used to develop biodegradable dust suppressants from biomass-based materials. This research highlights the solution′s role in reducing airborne particulate matter and improving environmental health (Oh et al., 2024).

  • Sustainability in Organic Waste Conversion: The solution is integral in evaluating the sustainability of processes that convert the organic fraction of municipal solid waste into biofuel and other value-added products. Such studies are essential for advancing waste-to-energy technologies and ensuring environmental sustainability (Musharavati et al., 2024).

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service