Skip to Content
Merck
All Photos(1)

Key Documents

CRM40901

Supelco

(+)-Borneol solution

certified reference material, 2000 μg/mL in methanol, ampule of 1 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$247.00

$247.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$247.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$247.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable

application(s)

cannabis testing
cannabis testing

format

single component solution

storage temp.

2-8°C

SMILES string

O[C@@H]1C2(C(C(C1)CC2)(C)C)C

InChI

1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3/t7?,8-,10?/m0/s1

InChI key

DTGKSKDOIYIVQL-ZCUBBSJVSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service