Skip to Content
Merck
All Photos(1)

Key Documents

CRM32016

Supelco

Triclopyr

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$130.00

$130.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$130.00

About This Item

Empirical Formula (Hill Notation):
C7H4Cl3NO3
CAS Number:
Molecular Weight:
256.47
Beilstein:
225301
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$130.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)COc1nc(Cl)c(Cl)cc1Cl

InChI

1S/C7H4Cl3NO3/c8-3-1-4(9)7(11-6(3)10)14-2-5(12)13/h1H,2H2,(H,12,13)

InChI key

REEQLXCGVXDJSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Application

Environmental
Triclopyr is a herbicide for perennial broad-leaved and woody weed control mainly on uncultivated areas, grassland, plantations and rangeland. As of Reg. (EC) No 1107/2009 triclopyr is approved for use as a plant protection product in the European Union (EU). Maximum residue levels (MRLs) have been set according to Reg. (EU) 2021/590 for triclopyr for various products of plant and animal origin in the range of 0.01-0.3 mg/kg. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service