Skip to Content
Merck
All Photos(1)

Key Documents

BP1270

Nevirapine

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Nevirapine, 11-Cyclopropyl-5,11-dihydro-4-methyl-6H-dipyrido[3,2-b:2′,3′-e][1,4]diazepin-6-one

Sign Into View Organizational & Contract Pricing

Select a Size

BP1270
$315.00

$315.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
BP1270
$315.00

About This Item

Empirical Formula (Hill Notation):
C15H14N4O
CAS Number:
Molecular Weight:
266.30
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$315.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

nevirapine

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

CC1=CC=NC2=C1NC(C(C=CC=N3)=C3N2C4CC4)=O

InChI

1S/C15H14N4O/c1-9-6-8-17-14-12(9)18-15(20)11-3-2-7-16-13(11)19(14)10-4-5-10/h2-3,6-8,10H,4-5H2,1H3,(H,18,20)

InChI key

NQDJXKOVJZTUJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Nevirapine BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Nevirapine Oral Suspension
  • Biochem/physiol Actions

    NNRTI (non-nucleoside reverse transcriptase inhibitor) of HIV-1
    Nevirapine is an allosteric, non-nucleoside inhibitor of HIV reverse transcriptase (NNRTI). The Ki for inhibition of wild-type RT by Nevirapine is 200 nM.

    Packaging

    Unit quantity: 200 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 2

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service