Skip to Content
Merck
All Photos(1)

Key Documents

BP1175

Fluconazole impurity standard

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Fluconazole, 2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$344.00

$344.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$344.00

About This Item

Empirical Formula (Hill Notation):
C13H12F2N6O
CAS Number:
Molecular Weight:
306.27
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$344.00


Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

fluconazole

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

mixture

storage temp.

2-8°C

SMILES string

FC1=CC(F)=C(C(CN2N=CN=C2)(O)CN3N=CN=C3)C=C1

InChI

1S/C13H12F2N6O/c14-10-1-2-11(12(15)3-10)13(22,4-20-8-16-6-18-20)5-21-9-17-7-19-21/h1-3,6-9,22H,4-5H2

InChI key

RFHAOTPXVQNOHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluconazole impurity standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Fluconazole Oral Suspension
  • Fluconazole Capsules
  • Fluconazole Infusion
  • Biochem/physiol Actions

    Fluconazole is an antifungal agent. It is highly selective inhibitor of fungal cytochrome P-450 sterol C-14 α-demethyllation. Fluconazole is a potent inhibitor of CYP2C9. Fluconazole interferes with fungal ergosterol synthesis and downregulates the metallothionein gene.
    Fluconazole is an antifungal agent; highly selctive inhibitor of fungal cytochrome P-450 sterol C-14 α-demethyllation. Potent inhibitor of CYP2C9. Interferes with fungal ergosterol synthesis; downregulates metallothionein gene.

    Packaging

    Unit quantity: 50 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    related product

    Product No.
    Description
    Pricing

    Pictograms

    Health hazardExclamation mark

    Signal Word

    Danger

    Hazard Statements

    Hazard Classifications

    Acute Tox. 4 Oral - Aquatic Chronic 3 - Lact. - Repr. 1B

    Storage Class Code

    6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Questions

    Reviews

    No rating value

    Active Filters

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service