Skip to Content
Merck
All Photos(1)

Key Documents

97719-U

Supelco

VersaFlash HTFP Station

extended length

Synonym(s):

VersaFlash Station with Extended Rods

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NC.05
Product 97719-U is not currently sold in your country. Contact Technical Service

technique(s)

LPLC: suitable

compatibility

for use with cartridges having an overall (stacked) length less than 425mm

Looking for similar products? Visit Product Comparison Guide

General description

The VersaFlash station secures the VersaPak cartridges for processing. The unique pre-compressed cartridge design eliminates the need for compression barrels making cartridge change-out fast and easy. Installing, removing, and changing out VersaPak cartridges in the station is fast and easy by simply turning and pulling out the handle. Scale-up to larger cartridges is just as easy by simply loosening the knob on the upper platen and adjusting it to the proper height for the larger or smaller cartridge. Most flash chromatography systems typically limit users to a narrow sample range, requiring additional hardware purchase and laborious change-out in order to work with larger sample sizes.

Legal Information

VersaFlash is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service