Skip to Content
Merck
All Photos(1)

Key Documents

96625

Supelco

3-Methyl-1,5-pentanediol

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$101.00

$101.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$101.00

About This Item

Linear Formula:
HOCH2CH2CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1697331
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

$101.00


Estimated to ship on26 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

technique(s)

HPLC: suitable

refractive index

n20/D 1.454

density

0.974 g/mL at 20 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CC(CCO)CCO

InChI

1S/C6H14O2/c1-6(2-4-7)3-5-8/h6-8H,2-5H2,1H3

InChI key

SXFJDZNJHVPHPH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

289.4 °F - closed cup

Flash Point(C)

143 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service