95934
2,6-Naphthalenedicarboxylic acid
analytical standard
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥95.0% (GC)
technique(s)
HPLC: suitable
mp
>300 °C (lit.)
format
neat
storage temp.
2-30°C
SMILES string
OC(=O)c1ccc2cc(ccc2c1)C(O)=O
InChI
1S/C12H8O4/c13-11(14)9-3-1-7-5-10(12(15)16)4-2-8(7)6-9/h1-6H,(H,13,14)(H,15,16)
InChI key
RXOHFPCZGPKIRD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service