Skip to Content
Merck
All Photos(1)

Key Documents

95929

Supelco

N,N′-Diphenylthiourea

analytical standard

Synonym(s):

1,3-Diphenyl-2-thiourea, Diphenylsulfourea, Thiocarbanilide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$101.00

$101.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$101.00

About This Item

Linear Formula:
(C6H5NH)2CS
CAS Number:
Molecular Weight:
228.31
Beilstein:
644277
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21

$101.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

technique(s)

HPLC: suitable

mp

152-155 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

S=C(Nc1ccccc1)Nc2ccccc2

InChI

1S/C13H12N2S/c16-13(14-11-7-3-1-4-8-11)15-12-9-5-2-6-10-12/h1-10H,(H2,14,15,16)

InChI key

FCSHMCFRCYZTRQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service