Skip to Content
Merck
All Photos(1)

Key Documents

95658

Supelco

Allyl methacrylate

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$101.00

$101.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$101.00

About This Item

Linear Formula:
CH2=C(CH3)COOCH2CH=CH2
CAS Number:
Molecular Weight:
126.15
Beilstein:
1747406
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.21

$101.00


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor pressure

4.6 mmHg ( 20 °C)

contains

20-120 ppm MEHQ as inhibitor

technique(s)

HPLC: suitable

refractive index

n20/D 1.436 (lit.)

bp

59-61 °C/43 mmHg (lit.)

density

0.938 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCC=C

InChI

1S/C7H10O2/c1-4-5-9-7(8)6(2)3/h4H,1-2,5H2,3H3

InChI key

FBCQUCJYYPMKRO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - STOT RE 2 Oral

Target Organs

Liver

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

94.1 °F - closed cup

Flash Point(C)

34.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service