Skip to Content
Merck
All Photos(1)

Key Documents

93884

Supelco

2-Amino-5-methylphenol

analytical standard

Synonym(s):

2-Hydroxy-4-methylaniline, 4-Amino-3-hydroxytoluene, 6-Amino-m-cresol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$99.70

$99.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$99.70

About This Item

Linear Formula:
H2NC6H3(CH3)OH
CAS Number:
Molecular Weight:
123.15
Beilstein:
386144
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

$99.70


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

159-162 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

Cc1ccc(N)c(O)c1

InChI

1S/C7H9NO/c1-5-2-3-6(8)7(9)4-5/h2-4,9H,8H2,1H3

InChI key

HCPJEHJGFKWRFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service