Skip to Content
Merck
All Photos(1)

Key Documents

93687

Supelco

Lithium bromide solution

certified reference material, TraceCERT®, Internal Standard for MARGA, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
$265.00

$265.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
2.5 L
$265.00

About This Item

Linear Formula:
LiBr
CAS Number:
Molecular Weight:
86.85
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.24

$265.00


Estimated to ship on30 May 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

4 mg/L in H2O

technique(s)

ion chromatography: suitable

SMILES string

[Li+].[Br-]

InChI

1S/BrH.Li/h1H;/q;+1/p-1

InChI key

AMXOYNBUYSYVKV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

High-precision argentometric titration was used for the determination of the bromide content in a Lithium bromide stock solution. The certified value of the Lithium content (mg/kg) is determined using high-performance inductively coupled plasma optical emission spectrometry (HP-ICP-OES). All measurements are traced gravimetrically to an internationally accepted reference material from NIST (USA). The values obtained from titration and HP-ICP-OES were checked for agreement. The values must agree with each other in the range of their uncertainty.

Application

This product is suitable for the Metrohm MARGA instruments as internal standard solution.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service