Skip to Content
Merck
All Photos(1)

Key Documents

90558

Supelco

N,N-Diethyl-p-phenylenediamine

analytical standard

Synonym(s):

4-Amino-N,N-diethylaniline

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$99.70

$99.70


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$99.70

About This Item

Linear Formula:
(C2H5)2NC6H4NH2
CAS Number:
Molecular Weight:
164.25
Beilstein:
879361
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

$99.70


Estimated to ship on01 June 2025


Request a Bulk Order

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.571 (lit.)

bp

115-116 °C/5 mmHg (lit.)

mp

19-21 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CCN(CC)c1ccc(N)cc1

InChI

1S/C10H16N2/c1-3-12(4-2)10-7-5-9(11)6-8-10/h5-8H,3-4,11H2,1-2H3

InChI key

QNGVNLMMEQUVQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

282.2 °F - closed cup

Flash Point(C)

139 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service