Skip to Content
Merck
All Photos(1)

Documents

90128R

Supelco

Aroclor 1254 solution

1000 μg/mL in hexane, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

packaging

ampule of 1 mL

concentration

1000 μg/mL in hexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

room temp

InChI

1S/C12H5Cl5/c13-8-3-1-2-6(10(8)15)7-4-5-9(14)12(17)11(7)16/h1-5H

InChI key

AUGNBQPSMWGAJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Te-Hao Chen et al.
Ecotoxicology and environmental safety, 75(1), 207-212 (2011-09-23)
Polychlorinated biphenyls (PCBs) are a group of widespread contaminants, and accumulation of PCBs has been observed in corals in the field. However, the toxic effects of PCBs on corals have not been investigated. In this study, we tested short and
Kyung Ho Kim et al.
Neurotoxicology, 32(6), 981-985 (2011-05-17)
Ortho-substituted polychlorinated biphenyls (PCBs) are a concern to human developmental health. Rat dams were exposed to an environmentally relevant mixture of PCBs, Aroclor 1254, or pure congener PCB 95 (6 mg/kg/day) during the perinatal period (GD 5 through PD 21).
Lidia Strużyńska et al.
Toxicology, 300(1-2), 12-18 (2012-05-26)
Aroclor 1254, a commercially produced mixture of polychlorinated biphenyls, is known to cause many adverse conditions, including neurotoxicity. It has been recently postulated that upregulation of N-methyl-d-aspartate receptors (NMDARs) and enhanced glutamate signalling which leads to excitotoxicity, is the mechanism
L A Martin et al.
Drug metabolism and disposition: the biological fate of chemicals, 40(3), 588-595 (2011-12-22)
Polychlorinated biphenyl (PCB) congeners differentially reduce serum thyroxine (T(4)) in rats, but little is known about their ability to affect biliary excretion of T(4). Thus, male Sprague-Dawley rats were orally administered Aroclor-1254, Aroclor-1242 (32 mg/kg per day), PCB-95, PCB-99, PCB-118
Jing An et al.
Toxicology, 295(1-3), 8-14 (2012-03-13)
Polychlorinated biphenyls (PCBs) are still prevalent in the environment despite the fact that they have been banned in many countries for several decades. Recent epidemiologic studies have demonstrated a link between PCBs exposure and pathological alterations of bone tissues. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service