Skip to Content
Merck
All Photos(1)

Key Documents

861222

Supelco

3,3-Dichlorobenzidine (2nd Lot)

certified reference material, 5000 μg/mL in methanol

Synonym(s):

3,3′-Dichlorobenzidine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10Cl2N2
CAS Number:
Molecular Weight:
253.13
UNSPSC Code:
12181503

grade

certified reference material
TraceCERT®

product line

TraceCERT®

feature

standard type calibration

packaging

ampule of 2 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C12H10Cl2N2/c13-9-5-7(1-3-11(9)15)8-2-4-12(16)10(14)6-8/h1-6H,15-16H2

InChI key

HUWXDEQWWKGHRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3,3'-Dichlorobenzidine and 3,3'-dichlorobenzidine dihydrochloride.
Report on carcinogens : carcinogen profiles, 11, III87-III89 (2004-01-01)
Kristian F Knoell et al.
Journal of toxicology and environmental health. Part A, 75(8-10), 551-556 (2012-06-13)
In a chemical plant in Germany producing 3,3'-dichlorobenzidine dihydrochloride for the manufacture of colorants, blood and urine samples were taken for biological monitoring. 3,3'-Dichlorobenzidine (DBZ) was analyzed in urine by thin-layer chromatography and subsequently further combined with analysis of adducts
M C Nyman et al.
Journal of the American Society for Mass Spectrometry, 10(11), 1152-1156 (1999-10-28)
3,3'-Dichlorobenzidine (DCB) and its degradation products, 3-chlorobenzidine (MCB) and benzidine, are of environmental concern because of their carcinogenic nature. The suitability of a small Fourier transform ion cyclotron resonance (FT-ICR) mass spectrometer for the analysis of these environmental contaminants in
3,3'-Dichlorobenzidine and 3,3'-dichlorobenzidine dihydrochloride.
Report on carcinogens : carcinogen profiles, 10, 85-87 (2004-08-25)
Jin-Heon Lee et al.
Industrial health, 41(3), 242-248 (2003-08-15)
3,3'-dichlorobenzidine (DCB) is suspected to be arcinogenic in experimental animal and human. Several studies have investigated excretion of metabolites in urine, hemoglobin adduction and cancer incidence among workers exposed occupationally to DCB. In these researches, metabolites of DCB had a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service