Skip to Content
Merck
All Photos(1)

Documents

843380

TSKgel® IEXPAK PW Bulk Media Sampler

phase various, DEAE-5PW, SP-5PW, SuperQ-5PW, bottle of 3 × 25 mL, 20 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

product line

TSKgel®

form

slurry

packaging

bottle of 3 × 25 mL

manufacturer/tradename

Tosoh 43380

parameter

20 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

particle size

20 μm

operating pH

2-13

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

LabPak products are multi-milliliter containers of TOYOPEARL and TSKgel bulk media products. Typically they contain 3 or 4 different ligand types offered for a particular chromatography mode. They are useful for developmental engineers who wish to familiarize themselves with resin physical properties in different buffer systems. The larger resin amounts in LabPak products allow the packing of wider ID and longer columns than available in the ToyoScreen products. This helps the developmental chemist or engineer to better measure under actual packing conditions such properties as dynamic binding capacity, selectivity and column efficiency.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

103.1 °F

Flash Point(C)

39.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service