Skip to Content
Merck
All Photos(1)

Key Documents

819314

TSKgel® SuperHZ (3 µm) HPLC Columns

L x I.D. 2 cm × 4.6 mm, polystyrene-divinylbenzene phase

Synonym(s):

TSKgel® Size Exclusion (H-Type) HPLC Guard Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501
Product 819314 is not currently sold in your country. Contact Technical Service

Product Name

TSKgel® SuperHZ Guard Column, phase polystyrene-divinylbenzene, L × I.D. 2 cm × 4.6 mm, 3 μm particle size

material

stainless steel

Agency

suitable for USP L21

product line

TSKgel®

manufacturer/tradename

Tosoh SuperHZ-L, 19314

parameter

0.4 mL/min flow rate
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

2 cm × 4.6 mm

matrix

polymer particle platform

matrix active group

polystyrene-divinylbenzene phase

particle size

3 μm

pore size

mixed

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Guard Column for 4.6mmID TSKgel SuperHZ1000-4000 and HZM-N & -M columns, 3μm
TSKgel Guard Columns are stand-alone and do not require separate holders.

Physical form

THF can be replaced with benzene, chloroform, toluene, xylene, dichloromethane, dichloroethane.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service