Skip to Content
Merck
All Photos(1)

Key Documents

814013

TSKgel® Ether-5PW (10 µm) HPLC Columns

L x I.D. 5 cm × 5 mm, ether phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
Product 814013 is not currently sold in your country. Contact Technical Service

Product Name

TSKgel® Ether-5PW HPLC Column, phase ether, L × I.D. 5 cm × 5 mm, 10 μm particle size

material

glass

product line

TSKgel®

manufacturer/tradename

Tosoh 14013

parameter

≤4 M salt concentration
≤50% organic solvent
1.0 mL/min flow rate
10-50 °C temperature
20 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

5 cm × 5 mm

matrix

hydroxylated methacrylic polymer

matrix active group

ether phase

particle size

10 μm

operating pH

2-12

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

Both hydrophobic interaction chromatography (HIC) and reversed-phase liquid chromatography separate on the basis of protein hydrophobicity and allow selective binding and desorption of proteins. However, HIC operates at significantly lower binding energy and uses aqueous mobile phases. These characteristics provide a gentle technique that is less likely to disturb protein conformation. As a result, HIC generally provides better activity recovery.
Three resin-based TSKgel HIC columns provide a range of hydrophobicities for chromatographic optimization. TSKgel Phenyl-5PW columns are most hydrophobic, followed by Ether-5PW, while the surface of Butyl-NPR columns is most polar or least hydrophobic. Ammonium sulfate concentration can be minimized by using a more hydrophobic packing such as Phenyl-5PW. We recommend the more polar Ether-5PW column for purifying more hydrophobic proteins. Ether-5PW and Phenyl-5PW packings are based on TSKgel G5000PW resin - 10 micron particles with 1000  pores. Butyl-NPR packing is prepared from 2.5 micron nonporous particles, allowing for rapid analyses and semi-micropreparative applications. All three column types can be cleaned with 0.2 M NaOH. Scale-up can be performed by using corresponding Toyopearl bulk resins.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service