Skip to Content
Merck
All Photos(1)

Key Documents

807477

TOYOPEARL® Butyl-650M Bulk Media

matrix polymer (65 μm), bottle of 100 mL, 65 μm

Synonym(s):

TOYOPEARL® Butyl-650 HIC Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
Product 807477 is not currently sold in your country. Contact Technical Service

description

Butyl-650M

product line

TOYOPEARL®

form

slurry

packaging

bottle of 100 mL

parameter

3 bar max. pressure

technique(s)

HPLC: suitable

matrix

polymer (65 μm)

matrix active group

C4 phase

particle size

65 μm

operating pH

1-13

capacity

40 g/L±10 adsorption capacity (lysozyme)

separation technique

hydrophobic interaction (HIC)

General description

TOYOPEARL® HIC resins are hydrophobic interaction chromatography resins that offer the following advantages: strong affinity for water-soluble proteins, high recovery of mass and activity, high sample capacity up to 2-4 times that of gel media, fluctuating salt concentrations will not change the bed volume, mechanical stability to 7kg/cm squared (7 bar/100psi), stability at a wide pH range (2-12), clean in place with 0.5M NaOH, and autoclavable. The exclusion limit of 5x106 Da and large pore size, 1000Å, enables these packings to separate very large proteins by a hydrophobic interaction mechanism, without size exclusion effects.
TOYOPEARL® HIC resins for hydrophobic interaction chromatography offer these advantages:
  • Strong affinity for water-soluble proteins
  • High recovery of mass and activity
  • High sample capacity: up to 2-4 times that of gel media
  • Fluctuating salt concentrations will not change bed volume
  • Mechanical stability to 7kg/cm2 (7 bar/100psi)
  • Stable at wide pH range: 2-12
  • Clean in place with 0.5M NaOH
  • Autoclavable
     
TOYOPEARL media offer high yield recovery of proteins, using various aqueous eluants. The exclusion limit of 5x106 Da and large pore size, 1000Å, enables these packings to separate very large proteins by a hydrophobic interaction mechanism, without size exclusion effects. Packed as 100mL bottle.

Application

TOYOPEARL® media are used in hydrophobic interaction media, resins and separation media. TOYOPEARL® media offer high yield recovery of proteins, using various aqueous eluants.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol.

Other Notes

Toyopearl Butyl-650 resin has the second highest hydrophobicity of the HIC ligands offered by TBL. Its pore size is the largest of our three butyl resins. Primary applications are for the separation of proteins, their isoforms, and aggregate removal. It is typically used in interresinte purification and polishing steps

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongwei Luan et al.
Sheng wu gong cheng xue bao = Chinese journal of biotechnology, 24(5), 867-873 (2008-08-30)
A beta-D-xylosidase from Leifsonia shinshuensis DICP 16 was purified to apparent homogeneity using a combination of ammonium sulfate precipitation, DE 52 anion-exchange, Q-Sepharose Fast Flow anion-exchange, Toyopearl Butyl 650C hydrophobic-interaction and Sephacryl S-300 HR gel-permeation chromatography. The purified xylosidase consisted
P Ceccaroli et al.
Journal of chromatography. B, Biomedical sciences and applications, 702(1-2), 41-48 (1998-02-04)
Hydrophobic interaction chromatography (HIC) has been used extensively for the separation of proteins and peptides by elution using a descending salt gradient, with and without the use of detergents or denaturing agents. In this paper we compare different hydrophobic interaction
V Stocchi et al.
Journal of chromatography. A, 676(1), 51-63 (1994-07-29)
Hydrophobic interaction chromatography (HIC) has been employed extensively in the separation of proteins by elution using a descending salt gradient, with and without the use of detergents or denaturing agents. In this study, a new hydrophobic interaction chromatographic support, Toyopearl
Hydrophobic interaction chromatography selectivity changes among three stable proteins: conformation does not play a major role
Jones, T.T. and Fernandez, E.J.
Biotechnology and Bioengineering, 87, 288-299 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service