Skip to Content
Merck
All Photos(1)

Key Documents

79585

Supelco

Metolcarb

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

m-Tolyl methylcarbamate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$199.00

$199.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$199.00

About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
Beilstein:
2045865
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$199.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

SMILES string

CNC(=O)Oc1cccc(C)c1

InChI

1S/C9H11NO2/c1-7-4-3-5-8(6-7)12-9(11)10-2/h3-6H,1-2H3,(H,10,11)

InChI key

VOEYXMAFNDNNED-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Metolcarb is a systemic carbamate insecticide, widely used to control planthoppers, leafhoppers, and other sucking insects in rice, citrus mealybugs, onion thrips, Mediterranean fruit flies, pink bollworms, and cotton aphids. It acts by inhibiting the activity of the acetylcholinesterase (AChE) enzyme and blocking the hydrolysis of acetylcholine, causing hyperexcitation.
Metolcarb is not approved for use in the European Union (EU) as a plant protection agent, following the Regulation (EC) No. 1107/2009, repealing the Directive 91/414/EEC. But a default maximum residue limit (MRL) of 0.01 mg/kg is set for metolcarb according to Art 18(1)(b) Reg 396/2005.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Metolcarb CRM may also be used in method development and validation to analyze:
  • 126 pesticide residues in chicken samples using modified QuEChERS for sample pre-treatment and ultra-high-performance liquid chromatography combined with quadrupole time-of-flight mass spectrometry (UHPLC-QToF-MS) for quantification
  • 250 pesticide residues extracted by the QuEChERS method from 109 processed fruit samples using ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • 252 pesticides extracted from surface water samples by solid-phase extraction (SPE) using liquid chromatography quadrupole-orbitrap high-resolution tandem mass spectrometry
  • Trace residues of 300 pesticides from fruit and vegetable surfaces in 30 seconds by thermal desorption electrospray ionization and tandem mass spectrometry (TD-ESI/MS/MS)
  • 379 pesticides in human serum samples extracted by QuEChERS method using liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • 117 pesticide residues in different tea samples extracted by in-syringe dispersive solid-phase extraction (IS-d-SPE) using ultra-performance liquid chromatography Orbitrap high-resolution mass spectrometry (UPLC-Q-Orbitrap-MS)

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service