Skip to Content
Merck
All Photos(1)

Key Documents

78945

Supelco

3-Hydroxyadipic acid sodium salt

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$498.00

$498.00


Estimated to ship on02 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$498.00

About This Item

Empirical Formula (Hill Notation):
C6H10O5 · xNa+
CAS Number:
Molecular Weight:
162.14 (free acid basis)
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.24

$498.00


Estimated to ship on02 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service