Skip to Content
Merck
All Photos(1)

Key Documents

77531

Supelco

Tralkoxydim

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$110.00

$110.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$110.00

About This Item

Empirical Formula (Hill Notation):
C20H27NO3
CAS Number:
Molecular Weight:
329.43
Beilstein:
8577523
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$110.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

CCO\N=C(/CC)C1=C(O)CC(CC1=O)c2c(C)cc(C)cc2C

InChI

1S/C20H27NO3/c1-6-16(21-24-7-2)20-17(22)10-15(11-18(20)23)19-13(4)8-12(3)9-14(19)5/h8-9,15,22H,6-7,10-11H2,1-5H3/b21-16+

InChI key

DQFPEYARZIQXRM-LTGZKZEYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Tralkoxydim is a selective and systemic herbicide that belongs to the group of cyclohexanedione oximes. It is an acetyl CoA carboxylase (ACCase) inhibitor that prevents the incorporation of acetate into fatty acids and blocks the production of phospholipids used in building new membranes. Tralkoxydim is used for post-emergence control of annual grass weeds including Avena spp., Lolium spp., Setaria viridis, Phalaris spp., Alopecurus myosuroides, and Apera spica-venti in wheat and barley.
According to the Commission Implementing Regulation (EU) No 540/2011 of 25th May 2011, tralkoxydim is not approved for use in the European Union (EU) as a plant protection agent, following the Regulation (EC) No. 1107/2009. But default maximum residue levels of 0.01 mg/kg to 0.05 mg/kg have been set for the presence of tralkoxydim (measured as the sum of its constituent isomers) in products of plant and animal origin, according to the EU Reg. No 2016/1015.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Tralkoxydim CRM may also be used as given below:

  • Evaluation of 252 pesticides extracted from surface water samples by solid-phase extraction (SPE) using liquid chromatography quadrupole-orbitrap high-resolution tandem mass spectrometry
  • Extraction of 250 pesticide residues by QuEChERS method from 103 processed fruit samples and their analysis by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • Screening of 448 pesticide residues in 10 samples of fruits and vegetables by a semiautomated method based on ultrahigh-performance liquid chromatography coupled with electrospray ionization quadrupole-Orbitrap high-resolution mass spectrometry (UHPLC/ESI Q-Orbitrap)
  • Development of a semi-automated method for the analysis of 261 pesticides and 41 environmental contaminants in catfish muscle samples by QuEChERS sample extraction and low-pressure GC-MS/MS and UHPLC-MS/MS, following an automated robotic micro-SPE clean-up
  • Screening and quantification of 569 pesticides in honeysuckle samples by a combination of ultrahigh-performance liquid chromatography (UHPLC) and quadrupole-Orbitrap high-resolution mass spectrometry
  • Analysis of 260 pesticides in high water content fruit and vegetables by liquid chromatography-tandem mass spectrometry (LC-MS/MS)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service