Skip to Content
Merck
All Photos(1)

Key Documents

72661

Supelco

4-Nitrophenol sodium salt hydrate

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$110.00

$110.00


Check Cart for Availability


Select a Size

Change View
100 MG
$110.00

About This Item

Linear Formula:
O2NC6H4ONa · xH2O
CAS Number:
Molecular Weight:
161.09 (anhydrous basis)
MDL number:
UNSPSC Code:
41116107

$110.00


Check Cart for Availability

grade

analytical standard

Quality Level

Assay

≥99% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

2-30°C

SMILES string

O.[Na+].[O-]c1ccc(cc1)[N+]([O-])=O

InChI

1S/C6H5NO3.Na.H2O/c8-6-3-1-5(2-4-6)7(9)10;;/h1-4,8H;;1H2/q;+1;/p-1

InChI key

SFNXTNREEGTEJL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service