Skip to Content
Merck
All Photos(1)

Key Documents

72656

Supelco

N-Tigloylglycine

analytical standard

Synonym(s):

N-Tiglylglycine, N-[(2E)-2-Methyl-1-oxo-2-buten-1-yl]glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO3
CAS Number:
Molecular Weight:
157.17
Beilstein:
2206218
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(CNC(/C(C)=C/C)=O)=O

InChI

1S/C7H11NO3/c1-3-5(2)7(11)8-4-6(9)10/h3H,4H2,1-2H3,(H,8,11)(H,9,10)/b5-3+

InChI key

WRUSVQOKJIDBLP-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Tigloylglycine is an amino acid metabolite, reportedly identified as a urinary marker for characterizing tuberculosis treatment failure.

Application

N-Tigloylglycine may be used as an analytical reference standard for the quantification of the analyte in human and animal urine samples using gas chromatography coupled to mass spectrometry (GC-MS).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatography-mass spectrometry profiles of urinary organic acids in healthy captive cheetahs (Acinonyx jubatus)
Tordiffe WSA, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1049(2), 8-15 (2017)
Solid phase extraction procedure for urinary organic acid analysis by gas chromatography mass spectrometry
Liu A, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 806(2), 283-287 (2004)
Urinary metabolite markers characterizing tuberculosis treatment failure
Luis L, et al.
Metabolomics, 13(10), 124-124 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service