Skip to Content
Merck
All Photos(1)

Key Documents

67506-U

Supelco

BIOshell A400 Protein C18 (3.4 μm) HPLC Columns

L × I.D. 0.5 cm × 3 mm, HPLC Guard Cartridge, pkg of 3 ea

Sign Into View Organizational & Contract Pricing

Select a Size

3 EA
$961.00

$961.00


Check Cart for Availability


Select a Size

Change View
3 EA
$961.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$961.00


Check Cart for Availability

Product Name

BIOshell A400 Protein C18 Guard Column, L × I.D. 0.5 cm × 3 mm, 3.4 μm particle size, pkg of 3 ea

material

stainless steel hardware

Quality Level

Agency

suitable for USP L1

product line

BIOshell

feature

endcapped

analyte chemical class(es)

proteins

packaging

pkg of 3 ea

manufacturer/tradename

BIOshell

extent of labeling

1% carbon loading

parameter

0-90 °C temperature
600 bar pressure (8702 psi)

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

L × I.D.

0.5 cm × 3 mm

surface area

15 m2/g

matrix

superficially porous particle

matrix active group

C18 (octadecyl), diisobutyloctadecyl bonding phase

particle size

3.4 μm

pore size

400 Å

operating pH range

1-8

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service