Skip to Content
Merck
All Photos(1)

Key Documents

64815-U

Supelco

Condenser

size small

Synonym(s):

SMALL SOXHLET CONDENSER, EA

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$229.00

$229.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$229.00

About This Item

UNSPSC Code:
12352200
NACRES:
NB.21

$229.00


Check Cart for Availability

Request a Bulk Order

size

small

Agency

suitable for EPA OTM-45

application(s)

PFAS testing
food and beverages

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service