Skip to Content
Merck
All Photos(1)

Key Documents

61863-U

Proteomix® HIC Butyl-NP5 (5 µm) HPLC Columns

L × I.D. 10 mm × 4 mm

Synonym(s):

Proteomix® HIC Butyl-NP5, Guard Cartridge with Holder

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,010.00

$1,010.00


Check Cart for Availability


Select a Size

Change View
1 EA
$1,010.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$1,010.00


Check Cart for Availability

Product Name

Proteomix® HIC Butyl-NP5, Guard cartridge with holder, 5 μm, NP, L × I.D. 10 mm × 4 mm

material

stainless steel hardware

product line

Proteomix®

manufacturer/tradename

Sepax 431NP5-4001C

packaging

1 ea of

parameter

6000 psi max. pressure
80 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

10 mm × 4 mm

impurities

<10 ppm metals

matrix

polymeric
polystyrene/divinylbenzene (Spherical, highly cross-linked)

matrix active group

C4 (butyl) phase

particle size

5 μm

pore size

non-porous

operating pH range

2-12

separation technique

hydrophobic interaction (HIC)

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service