Skip to Content
Merck
All Photos(1)

Key Documents

59557

Supelco

SUPELCOSIL LC-CN (5 µm) HPLC Columns

L × I.D. 2 cm × 4 mm Supelguard Guard Cartridge pkg of 2 ea, Guard Cartridge holder required for use

Synonym(s):

Supelguard LC-CN 2CM KIT

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$478.00

$478.00


Check Cart for Availability


Select a Size

Change View
1 KIT
$478.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

$478.00


Check Cart for Availability

Product Name

SUPELCOSIL LC-CN Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm

Agency

suitable for USP L10

packaging

pkg of 1 kit

technique(s)

HPLC: suitable

L × I.D.

2 cm × 4 mm

matrix active group

cyano phase

particle size

5 μm

pore size

120 Å

application(s)

food and beverages

separation technique

hydrophilic interaction (HILIC)
normal phase
reversed phase

Looking for similar products? Visit Product Comparison Guide

Footnote

Kit includes one cartridge, a stand-alone holder, a piece of tubing, and 2 nuts and ferrules.

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Paczkowski et al.
Polish journal of pharmacology, 47(5), 429-434 (1995-09-01)
This paper describes a simple high-performance liquid chromatographic (HPLC) method for the determination of diltiazem and desacetyldiltiazem in human serum. After basic methyl-tert-butyl ether extraction and back-extraction with hydrochloric acid, the drug and its metabolite was injected into a Supelcosil
S Fogli et al.
Therapeutic drug monitoring, 21(3), 367-375 (1999-06-12)
A single high-performance liquid chromatography (HPLC) method, suitable for the analysis of daunorubicin, idarubicin, doxorubicin, epirubicin, and their 13-dihydro metabolites is validated in the present study. Preparation of plasma samples was performed by a first extraction of analytes with a
H Kataoka et al.
Journal of analytical toxicology, 24(4), 257-265 (2000-06-29)
A simple and rapid method for the determination of amphetamine, methamphetamine, and their 3,4-methylenedioxy derivatives in urine samples was developed using automated in-tube solid-phase microextraction (SPME) coupled with liquid chromatography-electrospray ionization mass spectrometry (LC-ESI-MS). In-tube SPME is an extraction technique
H Kataoka et al.
Journal of chromatography. B, Biomedical sciences and applications, 731(2), 353-359 (1999-10-08)
The technique of automated in-tube solid-phase microextraction (SPME) coupled with liquid chromatography-electrospray ionization mass spectrometry (LC-ESI-MS) was evaluated for the determination of ranitidine. In-tube SPME is an extraction technique for organic compounds in aqueous samples, in which analytes are extracted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service