Skip to Content
Merck
All Photos(1)

Documents

59201

Supelco

Postcolumn Reactor Components, Glass Beads

size 75 μm, acid-washed/silanized

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

packaging

pkg of 25 g

size

75 μm

Looking for similar products? Visit Product Comparison Guide

General description

The equipment needed to perform post-column reactions can be relatively simple. These components (acid-washed/silanized) enable easy and economic construction of desired systems. For small peak volumes, the use of a column (5 cm x 4.6 mm) filled with 75 μm beads is recommended.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service