Key Documents
58768
Coupling
1/4-28 for 1/16 in. O.D., bore diam. 0.043 in.
Synonym(s):
SSI Unions
About This Item
UNSPSC Code:
41115700
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
SSI
bore diam.
0.043 in.
fitting
1/4-28 for 1/16 in. O.D.
General description
Stainless steel, 1/4-28 threads
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
SSI is a trademark of Scientific Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)