Skip to Content
Merck
All Photos(1)

Documents

58318

Supelco

SUPELCOSIL LC-PAH (5 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

SUPELCOSIL LC-PAH HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm

Agency

suitable for USP L1

manufacturer/tradename

SUPELCOSIL

parameter

0-70 °C temperature
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

matrix

silica gel, spherical particle platform

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL LC-PAH columns were designed specifically for analyses of the priority pollutant PAHs listed in US EPA Method 610. 2.1 mm and 3.0 mm columns save solvent and improve sensitivity when sample mass is limited. 3 μm columns provide extremely rapid, highly efficient analyses, while retaining the durability of porous silicas. They are excellent and economical substitutes for 1.5 μm nonporous silicas.
 

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Ana Ballesteros-Gómez et al.
Analytical chemistry, 81(21), 9012-9020 (2009-09-30)
In this work, magnetic nanoparticles (MNPs) coated with hemimicelles of alkyl (C(10)-C(18)) carboxylates, which have traditionally been used to prepare ferrofluids for technological applications, were used for the first time to extract organic contaminants from environmental water samples. The nanoparticles
S Moret et al.
Zeitschrift fur Lebensmittel-Untersuchung und -Forschung, 201(4), 322-326 (1995-10-01)
This paper proposes a simple HPLC method for the determination of polycyclic aromatic hydrocarbons (PAHs) in water, wine and beer. Samples were purified by PAH collection in solid-phase extraction (SPE) and analysed by reversed-phase HPLC (Supelcosil LC-PAH column from Supelco).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service