Key Documents
58257
Rheodyne® Fittings
long nut for 1/16 in. tubing, pkg of 10 ea
Select a Size
10 EA
$111.00
$111.00
Available to ship on14 April 2025Details
Request a Bulk Order
Select a Size
Change View
10 EA
$111.00
About This Item
UNSPSC Code:
41115700
$111.00
Available to ship on14 April 2025Details
Request a Bulk Order
material
stainless steel nut
description
long nut
packaging
pkg of 10 ea
fitting
long nut for 1/16 in. tubing
General description
The long Rheodyne nut has a longer hex portion for easier wrench access.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1
1 of 1
Questions
  1. Hi. I have a few question about product#58257 (long nut Rheodyne Fittings). Do these come with ferrules? If they don't, can you suggest a ferrule that suits these nuts? What size is the hex nut?
    1 answer
    1. This product includes the nut only. Product 58258 is the 1/16” ferrule needed. The hex nut is 1/4 inch. Please see the link below to review the recommended ferrule product:
      https://www.sigmaaldrich.com/US/en/product/supelco/58258
      Helpful?